Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcgf2
Entrez Gene ID
22658
Gene Name
polycomb group ring finger 2
Gene Aliases
Mel18, Rnf110, Zfp144, mel-18
Chromosome Location
Chr.11: 97688821 - 97701344 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163307.1 NP_001156779.1 4 - 5 506 84
NM_001163308.1 NP_001156780.1 3 - 4 368 84
NM_009545.2 NP_033571.1 6 - 7 773 84
GenBank mRNA AF483502.1 - 2 - 3 214 84
AF483503.1 - 2 - 3 214 84
AK142837.1 - 3 - 4 281 84
BC016419.1 - 4 - 5 460 84
BI991588.1 - 3 - 4 434 84
D90085.1 - 6 - 7 621 84

View Full Product Details