The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163307.1 | NP_001156779.1 | 10 - 11 | 949 | 74 |
NM_001163308.1 | NP_001156780.1 | 9 - 10 | 811 | 74 | |
NM_009545.2 | NP_033571.1 | 12 - 13 | 1216 | 74 | |
GenBank mRNA | AF483502.1 | - | 8 - 9 | 657 | 74 |
AF483503.1 | - | 8 - 9 | 657 | 74 | |
AK142837.1 | - | 9 - 10 | 724 | 74 | |
BC016419.1 | - | 10 - 11 | 903 | 74 | |
D90085.1 | - | 12 - 13 | 1064 | 74 |