The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103171.1 | NP_001096641.1 | 13 - 14 | 4837 | 88 |
NM_010862.2 | NP_034992.2 | 13 - 14 | 4837 | 88 | |
NM_182698.2 | NP_874357.2 | 12 - 13 | 1075 | 88 | |
GenBank mRNA | AB014510.1 | - | 9 - 10 | 840 | 88 |
AF053130.1 | - | 11 - 12 | 1043 | 88 | |
AF144095.1 | - | 13 - 14 | 4837 | 88 | |
AY331132.1 | - | 13 - 14 | 4837 | 88 | |
AY331133.1 | - | 12 - 13 | 1075 | 88 |