The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019763.2 | NP_062737.2 | 6 - 7 | 1750 | 75 |
GenBank mRNA | AB055980.1 | - | 5 - 6 | 1192 | 75 |
AF156529.1 | - | 5 - 6 | 1252 | 75 | |
AK137647.1 | - | 6 - 7 | 1638 | 75 |