Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sphk1
- Entrez Gene ID
-
20698
- Gene Name
- sphingosine kinase 1
- Gene Aliases
-
1110006G24Rik, Sk1, Spk1
- Chromosome Location
-
Chr.11: 116529817 - 116536675 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK028667.1 |
- |
1 - 1 |
202 |
81 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK003499.1 |
- |
|
|
|
Chr. - - [-] on Build |
81 |
GenBankmRNA |
AK009121.1 |
- |
|
|
|
Chr. - - [-] on Build |
81 |
View Full Product Details