The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026678.4 | NP_080954.4 | 6 - 7 | 652 | 73 |
GenBank mRNA | AK002231.1 | - | 5 - 6 | 591 | 73 |
AK010847.1 | - | 6 - 7 | 607 | 73 | |
AK172620.1 | - | 6 - 7 | 617 | 73 | |
BC052146.1 | - | 6 - 7 | 642 | 73 | |
CA983696.1 | - | 4 - 5 | 387 | 73 |