Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Erp44
Entrez Gene ID
76299
Gene Name
endoplasmic reticulum protein 44
Gene Aliases
1110001E24Rik, AI849526, AL033348, Txndc4
Chromosome Location
Chr.4: 48193331 - 48279601 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029572.2 NP_083848.1 2 - 3 275 135
GenBank mRNA AK003217.1 - 2 - 3 244 135
AK151497.1 - 2 - 3 276 135
AK160113.1 - 2 - 3 258 135
AK172973.1 - - 217 135
BC019558.1 - 2 - 3 182 135

View Full Product Details