Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nek8
Entrez Gene ID
140859
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 8
Gene Aliases
4632401F23Rik, b2b1449Clo, jck
Chromosome Location
Chr.11: 78166106 - 78176695 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080849.3 NP_543125.1 12 - 13 1807 73
GenBank mRNA AF407579.1 - 12 - 13 1757 73
AK014546.1 - 10 - 11 1983 73
AK154358.1 - 12 - 13 1770 73
BC057995.1 - 11 - 12 1889 73
BC070457.1 - 12 - 13 1756 73

View Full Product Details