Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Papln
Entrez Gene ID
170721
Gene Name
papilin, proteoglycan-like sulfated glycoprotein
Gene Aliases
E030033C16Rik
Chromosome Location
Chr.12: 83762992 - 83794875 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205343.1 NP_001192272.1 20 - 21 2861 56
NM_130887.3 NP_570957.2 19 - 20 2795 56
GenBank mRNA AF314171.1 - 19 - 20 2768 56
AK053198.1 - 20 - 21 2863 56
AK086767.1 - 19 - 20 2777 56
BC005747.1 - 1 - 2 332 56
BC132475.1 - 19 - 20 2735 56
BC137854.1 - 19 - 20 2735 56
BC144975.1 - 20 - 21 2801 56

View Full Product Details