The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167991.1 | NP_001161463.1 | 9 - 10 | 876 | 72 |
NM_133255.2 | NP_573518.2 | 9 - 10 | 876 | 72 | |
GenBank mRNA | AK030599.1 | - | 9 - 10 | 869 | 72 |
AK042048.1 | - | 9 - 10 | 864 | 72 | |
AK136069.1 | - | 9 - 10 | 870 | 72 | |
AK145768.1 | - | 9 - 10 | 844 | 72 | |
AK161612.1 | - | 9 - 10 | 829 | 72 | |
AK166954.1 | - | 9 - 10 | 856 | 72 | |
AK169073.1 | - | 9 - 10 | 844 | 72 | |
BC002226.1 | - | 1 - 2 | 85 | 72 | |
BC019486.1 | - | 9 - 10 | 852 | 72 | |
BC029967.1 | - | 9 - 10 | 817 | 72 | |
BC055881.1 | - | 9 - 10 | 822 | 72 | |
BC080744.1 | - | 9 - 10 | 866 | 72 |