Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asl
Entrez Gene ID
109900
Gene Name
argininosuccinate lyase
Gene Aliases
2510006M18Rik
Chromosome Location
Chr.5: 130011495 - 130024331 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133768.4 NP_598529.1 2 - 3 92 51
GenBank mRNA AK076453.1 - 2 - 3 107 51
AK138423.1 - 2 - 3 89 51
AK140160.1 - 2 - 3 107 51
AK148764.1 - 2 - 3 76 51
AK168739.1 - 2 - 3 87 51
BC016670.1 - 2 - 3 92 51

View Full Product Details