Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Moxd2
Entrez Gene ID
194357
Gene Name
monooxygenase, DBH-like 2
Gene Aliases
Dbhl1
Chromosome Location
Chr.6: 40878794 - 40887494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139296.2 NP_647457.2 2 - 3 433 101
GenBank mRNA AB065134.1 - 2 - 3 433 101

View Full Product Details