The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144526.3 | NP_653109.2 | 5 - 6 | 788 | 60 |
GenBank mRNA | AK011345.1 | - | 5 - 6 | 786 | 60 |
AK049710.1 | - | 5 - 6 | 789 | 60 | |
AK078429.1 | - | 5 - 6 | 789 | 60 | |
BC020115.1 | - | 5 - 6 | 774 | 60 | |
BC029115.1 | - | 5 - 6 | 774 | 60 |