The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146208.2 | NP_666320.1 | 2 - 3 | 430 | 98 |
GenBank mRNA | AB072931.1 | - | 2 - 3 | 297 | 98 |
AK031027.1 | - | 2 - 3 | 331 | 98 | |
BC034753.1 | - | 2 - 3 | 373 | 98 |