The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146208.2 | NP_666320.1 | 6 - 7 | 1030 | 75 |
GenBank mRNA | AB072931.1 | - | 6 - 7 | 897 | 75 |
BC024921.1 | - | 6 - 7 | 889 | 75 | |
BC034753.1 | - | 6 - 7 | 973 | 75 | |
DV646606.1 | - | 4 - 5 | 477 | 75 |