Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpne2
Entrez Gene ID
234577
Gene Name
copine II
Gene Aliases
3322401K10Rik
Chromosome Location
Chr.8: 94533028 - 94570529 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153507.2 NP_705727.1 9 - 10 1074 71
GenBank mRNA AK160238.1 - 9 - 10 1064 71
BC022128.1 - 9 - 10 1035 71
BC023348.1 - 4 - 5 372 71
BC031801.1 - 9 - 10 1074 71

View Full Product Details