The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153574.2 | NP_705802.1 | 5 - 6 | 638 | 62 |
GenBank mRNA | AJ510136.1 | - | 5 - 6 | 624 | 62 |
AK052421.1 | - | 5 - 6 | 579 | 62 | |
AK054311.1 | - | 5 - 6 | 625 | 62 | |
BC005554.1 | - | 5 - 6 | 543 | 62 | |
BC054053.1 | - | 5 - 6 | 612 | 62 | |
BC060978.1 | - | 5 - 6 | 575 | 62 | |
BC080861.1 | - | 5 - 6 | 638 | 62 |