Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mycbpap
Entrez Gene ID
104601
Gene Name
MYCBP associated protein
Gene Aliases
4932408B01Rik, AMAP-1, AW125474
Chromosome Location
Chr.11: 94501347 - 94532388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170671.2 NP_733771.2 15 - 16 2327 67
GenBank mRNA AK029929.1 - 14 - 15 2503 67
AK166780.1 - 8 - 9 1475 67
BC027327.1 - 11 - 12 1688 67
BC050808.1 - 4 - 5 850 67

View Full Product Details