The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178771.3 | NP_848886.1 | 1 - 2 | 98 | 87 |
GenBank mRNA | AK036321.1 | - | 1 - 2 | 106 | 87 |
AK041089.1 | - | 1 - 2 | 101 | 87 | |
AK048601.1 | - | 1 - 2 | 66 | 87 | |
AK049946.1 | - | 1 - 2 | 98 | 87 | |
AK163170.1 | - | 1 - 2 | 98 | 87 | |
BC025484.1 | - | 1 - 2 | 111 | 87 | |
BC098185.1 | - | 1 - 2 | 73 | 87 |