The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007923.2 | NP_031949.2 | 3 - 4 | 1421 | 62 |
GenBank mRNA | AK041964.1 | - | 1 - 2 | 103 | 62 |
AK156537.1 | - | 3 - 4 | 1418 | 62 | |
BC004798.1 | - | 1 - 2 | 666 | 62 | |
BC041679.1 | - | 1 - 2 | 654 | 62 | |
BC144844.1 | - | 3 - 4 | 1171 | 62 | |
BC145795.1 | - | 3 - 4 | 1171 | 62 | |
Z36885.1 | - | 4 - 5 | 1366 | 62 |