Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptprv
Entrez Gene ID
13924
Gene Name
protein tyrosine phosphatase, receptor type, V
Gene Aliases
Esp, OST, OST-PTP, mOST-PTP
Chromosome Location
Chr.1: 135108498 - 135132575 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007955.3 NP_031981.3 17 - 18 3754 93
GenBank mRNA AF300701.1 - 16 - 17 3231 93
AK156112.1 - 17 - 18 3757 93
U36488.1 - 13 - 14 2916 93

View Full Product Details