The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008226.2 | NP_032252.1 | 3 - 4 | 1176 | 64 |
GenBank mRNA | AB154248.1 | - | 3 - 4 | 1141 | 64 |
AF064873.1 | - | 3 - 4 | 695 | 64 | |
AJ225122.1 | - | 3 - 4 | 1176 | 64 |