The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008227.1 | NP_032253.1 | 1 - 2 | 369 | 95 |
GenBank mRNA | AJ225124.1 | - | 1 - 2 | 369 | 95 |
AK032225.1 | - | 1 - 2 | 374 | 95 | |
BC039156.1 | - | 1 - 2 | 313 | 95 | |
BC138319.1 | - | 1 - 2 | 343 | 95 | |
BC138321.1 | - | 1 - 2 | 343 | 95 |