The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010162.2 | NP_034292.2 | 10 - 11 | 2875 | 74 |
GenBank mRNA | AK132327.1 | - | 10 - 11 | 2852 | 74 |
BC004741.1 | - | 10 - 11 | 2239 | 74 | |
U78539.1 | - | 10 - 11 | 2379 | 74 | |
X96639.1 | - | 10 - 11 | 2646 | 74 |