The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010408.3 | NP_034538.2 | 3 - 4 | 1364 | 79 |
GenBank mRNA | AF028737.1 | - | 3 - 4 | 984 | 79 |
AJ225123.1 | - | 3 - 4 | 1204 | 79 |