Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Suv39h1
Entrez Gene ID
20937
Gene Name
suppressor of variegation 3-9 homolog 1 (Drosophila)
Gene Aliases
AI852103, AL022883, DXHXS7466e, H3-K9-HMTase 1, KMT1A, mIS6
Chromosome Location
Chr.X: 8061171 - 8074760 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011514.2 NP_035644.1 1 - 2 224 73
GenBank mRNA AF019969.1 - 1 - 2 50 73
AF193861.1 - 1 - 2 46 73
AK088405.1 - 1 - 2 41 73
AK132238.1 - 1 - 2 86 73
AK139757.1 - 1 - 2 193 73
AK169592.1 - 1 - 2 194 73
BC023860.1 - 1 - 2 101 73
BU053459.1 - 1 - 2 224 73

View Full Product Details