Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ets2
Entrez Gene ID
23872
Gene Name
E26 avian leukemia oncogene 2, 3' domain
Gene Aliases
AU022856, Ets-2
Chromosome Location
Chr.16: 95702407 - 95721049 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011809.3 NP_035939.3 4 - 5 541 91
GenBank mRNA AK038348.1 - 4 - 5 528 91
AK128933.1 - 4 - 5 493 91
AK143687.1 - 4 - 5 493 91
AK144974.1 - 4 - 5 492 91
AK145852.1 - 4 - 5 377 91
AK150175.1 - 4 - 5 471 91
AK151315.1 - 4 - 5 492 91
AK152750.1 - 4 - 5 495 91
AK153339.1 - 4 - 5 493 91
AK153478.1 - 4 - 5 493 91
BC005486.1 - 4 - 5 462 91
BC005504.1 - 4 - 5 402 91
CT010235.1 - 3 - 4 303 91
J04103.1 - 4 - 5 440 91

View Full Product Details