The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011809.3 | NP_035939.3 | 9 - 10 | 1431 | 89 |
GenBank mRNA | AK014602.2 | - | 9 - 10 | 1353 | 89 |
AK030537.1 | - | 5 - 6 | 733 | 89 | |
AK038348.1 | - | 9 - 10 | 1418 | 89 | |
AK128933.1 | - | 10 - 11 | 1305 | 89 | |
AK143687.1 | - | 9 - 10 | 1383 | 89 | |
AK145852.1 | - | 9 - 10 | 1267 | 89 | |
AK150175.1 | - | 9 - 10 | 1360 | 89 | |
AK151315.1 | - | 9 - 10 | 1382 | 89 | |
AK152750.1 | - | 9 - 10 | 1385 | 89 | |
AK153339.1 | - | 9 - 10 | 1383 | 89 | |
AK153478.1 | - | 9 - 10 | 1383 | 89 | |
BC005486.1 | - | 9 - 10 | 1352 | 89 | |
BC005504.1 | - | 9 - 10 | 1292 | 89 | |
CT010235.1 | - | 8 - 9 | 1193 | 89 | |
J04103.1 | - | 9 - 10 | 1330 | 89 |