Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stom
Entrez Gene ID
13830
Gene Name
stomatin
Gene Aliases
Epb7.2
Chromosome Location
Chr.2: 35313990 - 35337009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013515.2 NP_038543.1 3 - 4 326 63
GenBank mRNA AF093620.1 - 3 - 4 277 63
AK077435.1 - 3 - 4 294 63
AK139242.1 - 3 - 4 294 63
AK143724.1 - 3 - 4 294 63
AK148975.1 - 3 - 4 295 63
AK149689.1 - 3 - 4 294 63
AK149821.1 - 3 - 4 294 63
AK149991.1 - 3 - 4 294 63
AK151129.1 - 3 - 4 293 63
BC003789.1 - 3 - 4 289 63
U17297.1 - 3 - 4 326 63

View Full Product Details