The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013515.2 | NP_038543.1 | 6 - 7 | 751 | 95 |
GenBank mRNA | AF093620.1 | - | 6 - 7 | 702 | 95 |
AK077435.1 | - | 6 - 7 | 719 | 95 | |
AK139242.1 | - | 6 - 7 | 719 | 95 | |
AK143724.1 | - | 6 - 7 | 719 | 95 | |
AK148975.1 | - | 6 - 7 | 720 | 95 | |
AK149689.1 | - | 6 - 7 | 719 | 95 | |
AK149821.1 | - | 6 - 7 | 719 | 95 | |
AK149991.1 | - | 6 - 7 | 719 | 95 | |
AK151129.1 | - | 6 - 7 | 718 | 95 | |
U17297.1 | - | 6 - 7 | 751 | 95 |