Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ero1l
Entrez Gene ID
50527
Gene Name
ERO1-like (S. cerevisiae)
Gene Aliases
ERO1-L
Chromosome Location
Chr.14: 45283087 - 45318572 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015774.3 NP_056589.1 6 - 7 588 86
GenBank mRNA AK142595.1 - 6 - 7 579 86
AK159217.1 - 6 - 7 555 86
BC025102.1 - 6 - 7 540 86
CT010195.1 - 6 - 7 506 86
CT010259.1 - 6 - 7 506 86

View Full Product Details