The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016772.1 | NP_058052.1 | 3 - 4 | 364 | 61 |
GenBank mRNA | AF030343.1 | - | 3 - 4 | 364 | 61 |
AK088495.1 | - | 3 - 4 | 352 | 61 | |
BC068112.1 | - | 3 - 4 | 364 | 61 | |
BC087924.1 | - | 3 - 4 | 425 | 61 |