Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ech1
Entrez Gene ID
51798
Gene Name
enoyl coenzyme A hydratase 1, peroxisomal
Gene Aliases
AA617331
Chromosome Location
Chr.7: 28824804 - 28832248 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016772.1 NP_058052.1 6 - 7 606 81
GenBank mRNA AF030343.1 - 6 - 7 606 81
AK088495.1 - 6 - 7 594 81
BC068112.1 - 6 - 7 606 81
BC087924.1 - 6 - 7 667 81

View Full Product Details