Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hif3a
Entrez Gene ID
53417
Gene Name
hypoxia inducible factor 3, alpha subunit
Gene Aliases
Ipas, MOP7, NEPAS, bHLHe17
Chromosome Location
Chr.7: 17030993 - 17062427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162950.1 NP_001156422.1 2 - 3 261 96
NM_016868.3 NP_058564.2 2 - 3 292 96
GenBank mRNA AB289606.1 - 2 - 3 228 96
AF060194.1 - 2 - 3 292 96
BC120587.1 - 2 - 3 245 96

View Full Product Details