Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcirg1
Entrez Gene ID
27060
Gene Name
T cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3
Gene Aliases
ATP6N1C, ATP6a3, Atp6i, OC-116, OPTB1, Stv1, TIRC7, Vph1, oc
Chromosome Location
Chr.19: 3896050 - 3907133 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136091.2 NP_001129563.1 2 - 3 230 67
NM_001167784.1 NP_001161256.1 2 - 3 575 67
NM_016921.3 NP_058617.3 2 - 3 271 67
GenBank mRNA AB022322.1 - 2 - 3 137 67
AF218253.1 - 2 - 3 209 67
AK155140.1 - 2 - 3 576 67
AK189246.1 - 1 - 2 96 67
AK201621.1 - 1 - 2 96 67
BC006761.1 - 2 - 3 271 67
BC085234.1 - 2 - 3 131 67
CJ057829.1 - 2 - 3 231 67

View Full Product Details