Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif3d
Entrez Gene ID
55944
Gene Name
eukaryotic translation initiation factor 3, subunit D
Gene Aliases
66/67kDa, AA407891, Eif3s7, eIF3p66
Chromosome Location
Chr.15: 77958998 - 77971088 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018749.2 NP_061219.2 11 - 12 1223 77
GenBank mRNA AB012580.1 - 11 - 12 1164 77
AK045228.1 - 11 - 12 1165 77
AK049767.1 - 11 - 12 1162 77
AK145689.1 - 11 - 12 1178 77
AK168256.1 - 11 - 12 1213 77
BC018598.1 - 11 - 12 1161 77
BC089020.1 - 11 - 12 1157 77

View Full Product Details