The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018792.1 | NP_061262.1 | 1 - 1 | 750 | 74 |
GenBank mRNA | AB022320.1 | - | 1 - 1 | 750 | 74 |
AB045724.1 | - | 1 - 1 | 743 | 74 | |
AK005718.1 | - | 1 - 1 | 781 | 74 | |
AY286319.1 | - | 1 - 1 | 750 | 74 | |
BC050966.1 | - | 1 - 1 | 783 | 74 | |
BC061119.1 | - | 1 - 1 | 813 | 74 |