Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hhip
Entrez Gene ID
15245
Gene Name
Hedgehog-interacting protein
Gene Aliases
Hhip1
Chromosome Location
Chr.8: 79965850 - 80058070 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020259.4 NP_064655.4 4 - 5 1323 83
GenBank mRNA AF116865.1 - 4 - 5 1304 83
AK031539.1 - 4 - 5 1322 83
AK031841.1 - 4 - 5 833 83
BC053012.1 - 4 - 5 1308 83

View Full Product Details