Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eed
Entrez Gene ID
13626
Gene Name
embryonic ectoderm development
Gene Aliases
ENSMUSG00000039373, l(7)5Rn, l7Rn5, lusk
Chromosome Location
Chr.7: 89954654 - 89981419 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021876.3 NP_068676.1 6 - 7 1076 83
GenBank mRNA AK077664.1 - 6 - 7 661 83
AK131976.1 - 6 - 7 643 83
AK133890.1 - 6 - 7 637 83
BC012966.1 - 6 - 7 1075 83
U78103.1 - 6 - 7 1078 83
U97675.1 - 6 - 7 1083 83

View Full Product Details