The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022313.2 | NP_071708.2 | 9 - 10 | 1213 | 73 |
GenBank mRNA | AB049389.1 | - | 9 - 10 | 1189 | 73 |
AF190730.1 | - | 9 - 10 | 1181 | 73 | |
AF190731.1 | - | 8 - 9 | 949 | 73 | |
AK012155.1 | - | 9 - 10 | 1203 | 73 | |
BC019728.1 | - | 3 - 4 | 401 | 73 | |
BC037603.1 | - | 8 - 9 | 1315 | 73 | |
BC068271.1 | - | 9 - 10 | 1208 | 73 |