Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adck3
Entrez Gene ID
67426
Gene Name
aarF domain containing kinase 3
Gene Aliases
4632432J16Rik, AI462003, Cabc1, mKIAA0451
Chromosome Location
Chr.1: 180165238 - 180196020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163290.1 NP_001156762.1 10 - 11 1308 65
NM_023341.3 NP_075830.2 10 - 11 1406 65
GenBank mRNA AJ278735.1 - 10 - 11 1286 65
AK004177.1 - 3 - 4 291 65
AK004791.1 - 10 - 11 1348 65
AK014605.2 - 10 - 11 1348 65
AK191635.1 - 1 - 2 31 65
AK192575.1 - 1 - 2 31 65
AK194890.1 - 1 - 2 31 65
AK196005.1 - 1 - 2 31 65
AK202430.1 - 1 - 2 31 65
AK207786.1 - 1 - 2 31 65
AK217974.1 - 2 - 3 146 65
BC030937.1 - 10 - 11 1354 65
U31629.1 - 8 - 9 947 65

View Full Product Details