The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023395.2 | NP_075884.1 | 4 - 5 | 714 | 76 |
GenBank mRNA | AK009971.1 | - | 4 - 5 | 717 | 76 |
AK018575.1 | - | 4 - 5 | 575 | 76 | |
AK132541.1 | - | 4 - 5 | 715 | 76 | |
AK142707.1 | - | 4 - 5 | 583 | 76 | |
AK161606.1 | - | 4 - 5 | 580 | 76 | |
BC025850.1 | - | 4 - 5 | 527 | 76 | |
BC038122.1 | - | 4 - 5 | 556 | 76 | |
BC132073.1 | - | 4 - 5 | 643 | 76 | |
BC132469.1 | - | 4 - 5 | 643 | 76 | |
BC145305.1 | - | 4 - 5 | 640 | 76 |