The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023403.3 | NP_075892.3 | 1 - 2 | 472 | 102 |
GenBank mRNA | AK008735.1 | - | 1 - 2 | 189 | 102 |
AK031949.1 | - | 1 - 2 | 215 | 102 | |
AK076760.1 | - | 1 - 2 | 212 | 102 | |
AK076773.1 | - | 1 - 2 | 207 | 102 | |
BC014742.1 | - | 1 - 2 | 193 | 102 | |
BC058612.1 | - | - | 3126 | 102 | |
BU054434.1 | - | 1 - 2 | 472 | 102 |