The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023596.3 | NP_076085.1 | 3 - 4 | 433 | 66 |
GenBank mRNA | AF326986.1 | - | 3 - 4 | 444 | 66 |
AK034133.1 | - | 3 - 4 | 429 | 66 | |
AK051317.1 | - | 3 - 4 | 458 | 66 | |
AK157868.1 | - | 3 - 4 | 434 | 66 | |
BC137864.1 | - | 3 - 4 | 416 | 66 | |
BC144983.1 | - | 3 - 4 | 416 | 66 | |
CV557129.1 | - | 2 - 3 | 303 | 66 |