The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023605.2 | NP_076094.2 | 1 - 2 | 369 | 137 |
GenBank mRNA | AB041585.1 | - | 1 - 2 | 338 | 137 |
AK018482.1 | - | 1 - 2 | 320 | 137 | |
BC020074.1 | - | 1 - 2 | 340 | 137 |