The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023712.3 | NP_076201.2 | 11 - 12 | 2349 | 49 |
NR_045537.1 | 12 - 13 | 2328 | 49 | ||
GenBank mRNA | AF212372.1 | - | 11 - 12 | 1840 | 49 |
AK008721.1 | - | 12 - 13 | 1825 | 49 | |
AK033907.1 | - | 9 - 10 | 2904 | 49 | |
AK154353.1 | - | 11 - 12 | 1838 | 49 | |
AK166931.1 | - | 11 - 12 | 1841 | 49 | |
BC002297.1 | - | 11 - 12 | 1368 | 49 | |
BC026854.1 | - | 11 - 12 | 1810 | 49 | |
BC085491.1 | - | 11 - 12 | 1793 | 49 |