Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crot
Entrez Gene ID
74114
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
1200003H03Rik
Chromosome Location
Chr.5: 8966023 - 8997146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023733.3 NP_076222.1 4 - 5 400 93
GenBank mRNA AK004567.2 - 4 - 5 401 93
AK037973.1 - 3 - 4 852 93
BC006593.1 - 4 - 5 388 93
BC012308.1 - 4 - 5 415 93

View Full Product Details