Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mphosph8
Entrez Gene ID
75339
Gene Name
M-phase phosphoprotein 8
Gene Aliases
1500035L22Rik, 4930548G07Rik, AU040635
Chromosome Location
Chr.14: 56668240 - 56697430 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023773.2 NP_076262.2 10 - 11 2273 130
GenBank mRNA AB041598.2 - 10 - 11 2274 130
AK016067.1 - 5 - 6 518 130
AK148654.1 - 3 - 4 276 130
AK158773.1 - 10 - 11 2251 130
AK159495.1 - 10 - 11 2251 130
BC079558.1 - 10 - 11 2205 130
BC145700.1 - 10 - 11 2198 130

View Full Product Details