The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023788.3 | NP_076277.1 | 1 - 1 | 468 | 90 |
GenBank mRNA | AF319981.1 | - | 1 - 1 | 243 | 90 |
AK008349.1 | - | 1 - 1 | 102 | 90 | |
AK145866.1 | - | 1 - 1 | 430 | 90 | |
BC051935.1 | - | 1 - 1 | 257 | 90 | |
BC058380.1 | - | 1 - 1 | 247 | 90 | |
BC060080.1 | - | 1 - 1 | 257 | 90 | |
CD363769.1 | - | 1 - 1 | 468 | 90 |