The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023910.6 | NP_076399.4 | 3 - 4 | 1625 | 74 |
NM_029805.1 | NP_084081.1 | 3 - 4 | 1625 | 74 | |
GenBank mRNA | AF201286.1 | - | 3 - 4 | 1338 | 74 |
AK018735.2 | - | 3 - 4 | 1598 | 74 | |
AK040534.1 | - | 3 - 4 | 1562 | 74 | |
AK143608.1 | - | 3 - 4 | 1402 | 74 | |
AK144806.1 | - | 2 - 3 | 1062 | 74 | |
AK150043.1 | - | 3 - 4 | 1594 | 74 | |
AK154153.1 | - | 3 - 4 | 1575 | 74 | |
AK154490.1 | - | 3 - 4 | 1225 | 74 | |
AK155712.1 | - | 3 - 4 | 1627 | 74 | |
AK170487.1 | - | 3 - 4 | 1614 | 74 | |
BC018544.1 | - | 2 - 3 | 1097 | 74 | |
BC023761.1 | - | 3 - 4 | 1529 | 74 | |
BC145721.1 | - | 3 - 4 | 1588 | 74 | |
BC145723.1 | - | 3 - 4 | 1588 | 74 |